Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4S)-4-N-Fmoc-amino-1-Boc-L-proline 95%

Supplier: AMBEED, INC
Warning

A436286-100MG A436286-1G A436286-250MG A436286-25G A436286-5G
77143-314EA 15.45 USD InStock
77143-314 77143-316 77143-318 77143-320 77143-322
(4S)-4-N-Fmoc-amino-1-Boc-L-proline 95%
(4S)-4-N-Fmoc-amino-1-Boc-L-proline
Precaución: for Research Use Only


Formula: C₂₅H₂₈N₂O₆
MW: 452.49 g/mol
MDL Number: MFCD00673782
CAS Number: 174148-03-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty