Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-phenoxybenzoate 99%

Supplier: AMBEED, INC
Warning

A438866-100G A438866-25G A438866-5G
77143-356EA 316.21 USD InStock
77143-356 77143-358 77143-360
Methyl-2-phenoxybenzoate 99%
Methyl 2-phenoxybenzoate
Precaución: for Research Use Only


Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
MDL Number: MFCD03093064
CAS Number: 21905-56-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty