Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-Ethoxycyclopropoxy)trimethylsilane 97%

Supplier: AMBEED, INC
Danger

A440123-1G A440123-250MG A440123-5G A440123-25G
77143-372EA 31.93 USD InStock
77143-372 77143-374 77143-376 77551-322
(1-Ethoxycyclopropoxy)trimethylsilane 97%
(1-Ethoxycyclopropoxy)trimethylsilane
Precaución: for Research Use Only


Formula: C₈H₁₈O₂Si
MDL Number: MFCD00074986
CAS Number: 27374-25-0
UN: 1993
ADR: 3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty