Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

QNZ46 98%

Supplier: AMBEED, INC
Warning

A443937-10MG A443937-1MG A443937-50MG A443937-5MG
77143-414EA 9.27 USD InStock
77143-414 77143-416 77143-418 77143-420
QNZ46 98%
QNZ46
Precaución: for Research Use Only


Formula: C₂₄H₁₇N₃O₆
MDL Number: MFCD22666391
CAS Number: 1237744-13-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty