Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Fluoro-5-(methoxycarbonyl)-2-methylphenyl)boronic acid 97%

Supplier: AMBEED, INC
Warning

A471020-100MG A471020-250MG
77143-678EA 50.47 USD InStock
77143-678 77143-680
(3-Fluoro-5-(methoxycarbonyl)-2-methylphenyl)boronic acid 97%
(3-Fluoro-5-(methoxycarbonyl)-2-methylphenyl)boronic acid
Precaución: for Research Use Only


Formula: C₉H₁₀BFO₄
MW: 211.98 g/mol
MDL Number: MFCD28291813
CAS Number: 2828446-97-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty