Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-(4-(hydroxymethyl)phenyl)acetate 98%

Supplier: AMBEED, INC
Warning

A471739-100MG A471739-1G A471739-250MG A471739-5G A471739-25G
77143-682EA 13.39 USD InStock
77143-682 77143-684 77143-686 77143-688 77746-965
Methyl 2-(4-(hydroxymethyl)phenyl)acetate 98%
Methyl 2-(4-(hydroxymethyl)phenyl)acetate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD08061910
CAS Number: 155380-11-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty