Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-1-(2-Naphthyl)ethanol 95%

Supplier: AMBEED, INC
Warning

A476681-1G A476681-250MG A476681-5G
77143-748EA 36.04 USD InStock
77143-748 77143-750 77746-979
(R)-(+)-1-(2-Naphthyl)ethanol 95%
(R)-(+)-1-(2-Naphthyl)ethanol
Precaución: for Research Use Only


Formula: C₁₂H₁₂O
MDL Number: MFCD00075344
CAS Number: 52193-85-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty