Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(3-Methylphenyl)-1H-tetrazole 95%

Supplier: AMBEED, INC
Danger

A477837-1G A477837-250MG
77143-768EA 17.51 USD InStock
77143-768 77143-770
5-(3-Methylphenyl)-1H-tetrazole 95%
5-(3-Methylphenyl)-1H-tetrazole
Precaución: for Research Use Only


Formula: C₈H₈N₄
MDL Number: MFCD03093096
CAS Number: 3441-00-7
UN: 1325
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty