Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)benzenesulphonamide 98%

Supplier: AMBEED, INC
Warning

A480098-100G A480098-1G A480098-25G A480098-5G
77143-786EA 169.95 USD InStock
77143-786 77143-788 77143-790 77143-792
3-(Trifluoromethyl)benzenesulphonamide 98%
3-(Trifluoromethyl)benzenesulphonamide
Precaución: for Research Use Only


Formula: C₇H₆F₃NO₂S
MDL Number: MFCD00042421
CAS Number: 672-58-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty