Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-Phenylsuccinic acid 97%

Supplier: AMBEED, INC
Warning

A481229-1G A481229-250MG A481229-5G
77143-798EA 38.12 USD InStock
77143-798 77143-800 77143-802
(R)-(-)-Phenylsuccinic acid 97%
(R)-(-)-Phenylsuccinic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₄
MW: 194.19
Boiling Pt: 368
MDL Number: MFCD00065930
CAS Number: 46292-93-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty