Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromobenzhydrazide 98%

Supplier: AMBEED, INC
Warning

A481801-100G A481801-1G A481801-25G A481801-5G
77143-814EA 77.25 USD InStock
77143-814 77143-816 77143-818 77143-820
4-Bromobenzhydrazide 98%
4-Bromobenzhydrazide
Precaución: for Research Use Only


Formula: C₇H₇BrN₂O
MDL Number: MFCD00007602
CAS Number: 5933-32-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty