Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tin(II) bromide 99%

Supplier: AMBEED, INC
Danger

A487179-1G A487179-25G A487179-5G
77143-878EA 10.3 USD InStock
77143-878 77143-880 77143-882
Tin(II) bromide 99%
Tin(II) bromide
Precaución: for Research Use Only


Formula: Br₂Sn
MDL Number: MFCD00011239
CAS Number: 10031-24-0
UN: 3260
ADR: 8,Ⅱ
Order Now
Specification Test Results
Loss on drying ≤ 2%

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty