Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-3-hydroxybenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A506664-100MG A506664-1G A506664-250MG
77144-068EA 133.9 USD InStock
77144-068 77144-070 77144-072
2,6-Dichloro-3-hydroxybenzaldehyde 97%
2,6-Dichloro-3-hydroxybenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
MDL Number: MFCD24673790
CAS Number: 56962-14-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty