Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SW-033291 98%

Supplier: AMBEED, INC
Warning

A509020-10MG A509020-1MG A509020-25MG A509020-5MG
77144-084EA 54.59 USD InStock
77144-084 77144-086 77144-088 77144-090
SW-033291 98%
SW-033291
Precaución: for Research Use Only


Formula: C₂₁H₂₀N₂OS₃
MW: 412.59 g/mol
MDL Number: MFCD02973087
CAS Number: 459147-39-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty