Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Bromomethyl)cyclobutane 98%

Supplier: AMBEED, INC
Danger

A512608-1G A512608-25G A512608-10G
77144-112EA 9.26 USD InStock
77144-112 77551-538 77572-630
(Bromomethyl)cyclobutane 98%
(Bromomethyl)cyclobutane
Precaución: for Research Use Only


Formula: C₅H₉Br
MW: 149.03
Density: 1.35
Flash Pt: 41
MDL Number: MFCD00040960
CAS Number: 17247-58-4
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty