Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Bromophenoxy)-tert-butyldimethylsilane 95%

Supplier: AMBEED, INC
Danger

A525795-100G A525795-1G A525795-5G
77144-212EA 155.53 USD InStock
77144-212 77144-214 77144-216
(4-Bromophenoxy)-tert-butyldimethylsilane 95%
(4-Bromophenoxy)-tert-butyldimethylsilane
Precaución: for Research Use Only


Formula: C₁₂H₁₉BrOSi
MW: 287.272 g/mol
MDL Number: MFCD00239382
CAS Number: 67963-68-2
UN: 1993
ADR: 3 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty