Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-cyclopropylquinazoline 97%

Supplier: AMBEED, INC
Warning

A530595-100MG A530595-250MG A530595-1G
77144-268EA 15.45 USD InStock
77144-268 77686-372 77686-371
4-Chloro-2-cyclopropylquinazoline 97%
4-Chloro-2-cyclopropylquinazoline
Precaución: for Research Use Only


Formula: C₁₁H₉ClN₂
MDL Number: MFCD14453157
CAS Number: 898044-61-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty