Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-(((Benzyloxy)carbonyl)amino)-5-methylhexanoic acid 97%

Supplier: AMBEED, INC
Warning

A532460-100MG A532460-1G A532460-250MG
77144-280EA 20.6 USD InStock
77144-280 77144-282 77144-284
(S)-3-(((Benzyloxy)carbonyl)amino)-5-methylhexanoic acid 97%
(S)-3-(((Benzyloxy)carbonyl)amino)-5-methylhexanoic acid
Precaución: for Research Use Only


Formula: C₁₅H₂₁NO₄
MW: 279.33 g/mol
MDL Number: MFCD19382362
CAS Number: 118247-68-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty