Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Fluorophenyl)methanesulfonyl chloride 98%

Supplier: AMBEED, INC
Danger

A535935-1G A535935-250MG A535935-25G A535935-5G
77144-348EA 47.38 USD InStock
77144-348 77144-350 77144-352 77144-354
(4-Fluorophenyl)methanesulfonyl chloride 98%
(4-Fluorophenyl)methanesulphonyl chloride
Precaución: for Research Use Only


Formula: C₇H₆ClFO₂S
MW: 208.64 g/mol
MDL Number: MFCD01631930
CAS Number: 103360-04-9
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty