Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-cyano-2-fluorobenzoate 95%

Supplier: AMBEED, INC
Warning

A539756-1G A539756-250MG A539756-5G
77144-384EA 23.68 USD InStock
77144-384 77144-386 77144-388
Methyl 3-cyano-2-fluorobenzoate 95%
Methyl 3-cyano-2-fluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₆FNO₂
MDL Number: MFCD18399057
CAS Number: 1261453-50-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty