Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-N'-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)benzohydrazide 97%

Supplier: AMBEED, INC
Warning

A549010-1G A549010-250MG A549010-5G
77144-460EA 216.3 USD InStock
77144-460 77144-462 77144-464
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-N'-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)benzohydrazide 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-N'-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)benzohydrazide
Precaución: for Research Use Only


Formula: C₂₆H₃₄B₂N₂O₆
MDL Number: MFCD18914530
CAS Number: 2096338-52-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty