Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyanobenzenesulfonyl chloride 96%

Supplier: AMBEED, INC
Danger

A554287-1G A554287-25G A554287-5G
77144-518EA 12.36 USD InStock
77144-518 77144-520 77144-522
2-Cyanobenzenesulfonyl chloride 96%
2-Cyanobenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₇H₄ClNO₂S
MDL Number: MFCD00052341
CAS Number: 69360-26-5
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty