Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diiodo(p-cymene)ruthenium(II) dimer 98%

Supplier: AMBEED, INC
Warning

A568362-100MG A568362-1G A568362-250MG A568362-5G
77144-610EA 11.33 USD InStock
77144-610 77144-612 77144-614 77144-616
Diiodo(p-cymene)ruthenium(II) dimer 98%
Diiodo(p-cymene)ruthenium(II) dimer
Precaución: for Research Use Only


Formula: C₂₀H₂₈I₄Ru₂
MDL Number: MFCD08064027
CAS Number: 90614-07-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty