Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)anilinium chloride 98+%

Supplier: AMBEED, INC
Warning

A569874-25G A569874-5G A569874-1G
77144-626EA 77.25 USD InStock
77144-626 77144-628 77747-175
4-(Trifluoromethyl)anilinium chloride 98+%
4-(Trifluoromethyl)anilinium chloride
Precaución: for Research Use Only


Formula: C₇H₇ClF₃N
MW: 197.59
Melting Pt: 200
MDL Number: MFCD00007897
CAS Number: 90774-69-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty