Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Thien-3-ylacrylic acid 95%

Supplier: AMBEED, INC
Warning

A575483-100G A575483-10G A575483-1G A575483-25G A575483-5G
77144-676EA 489.25 USD InStock
77144-676 77144-678 77144-680 77144-682 77144-684
3-Thien-3-ylacrylic acid 95%
3-Thien-3-ylacrylic acid
Precaución: for Research Use Only


Formula: C₇H₆O₂S
MDL Number: MFCD00014535
CAS Number: 102696-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty