Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloro-4-pyridineboronic acid 95%

Supplier: AMBEED, INC
Warning

A576420-10G A576420-1G A576420-5G
77144-692EA 46.35 USD InStock
77144-692 77144-694 77144-696 77144-698
2,3-Dichloro-4-pyridineboronic acid 95%
2,3-Dichloro-4-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₅H₄BCl₂NO₂
MW: 191.81 g/mol
MDL Number: MFCD04038756
CAS Number: 951677-39-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty