Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 7-amino-5-azaspiro[2.4]heptane-5-carboxylate 98%

Supplier: AMBEED, INC
Danger

A577318-250MG A577318-1G
77144-700EA 52.53 USD InStock
77144-700 77686-467
tert-Butyl 7-amino-5-azaspiro[2.4]heptane-5-carboxylate 98%
tert-Butyl 7-amino-5-azaspiro[2.4]heptane-5-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD18375095
CAS Number: 1026609-83-5
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty