Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dioxopyrrolidin-1-yl 3-(2-((tert-butoxycarbonyl)amino)ethoxy)propanoate 98%

Supplier: AMBEED, INC
Warning

A581867-1G A581867-250MG A581867-100MG
77144-726EA 211.15 USD InStock
77144-726 77144-728 77686-480
2,5-Dioxopyrrolidin-1-yl 3-(2-((tert-butoxycarbonyl)amino)ethoxy)propanoate 98%
2,5-Dioxopyrrolidin-1-yl 3-(2-((tert-butoxycarbonyl)amino)ethoxy)propanoate
Precaución: for Research Use Only


Formula: C₁₄H₂₂N₂O₇
MDL Number: MFCD20227793
CAS Number: 1260092-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty