Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

E6005 (RVT-501) 98%

Supplier: AMBEED, INC
Warning

A585384-10MG A585384-1MG A585384-25MG A585384-5MG
77144-774EA 59.74 USD InStock
77144-774 77144-776 77144-778 77144-780
E6005 (RVT-501) 98%
E6005 (RVT-501)
Precaución: for Research Use Only


Formula: C₂₆H₂₄N₄O₅
MDL Number: MFCD28502071
CAS Number: 947620-48-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty