Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzyloxy-2-fluorophenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A609977-1G A609977-250MG A609977-5G
77144-954EA 89.61 USD InStock
77144-954 77144-956 77144-958
4-Benzyloxy-2-fluorophenylboronic acid pinacol ester 98%
4-Benzyloxy-2-fluorophenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₉H₂₂BFO₃
MDL Number: MFCD22419262
CAS Number: 1823170-17-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty