Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R,R)-(-)-2,5-Hexanediol 98%

Supplier: AMBEED, INC
Warning

A616579-100MG A616579-1G A616579-250MG A616579-5G
77145-002EA 15.45 USD InStock
77145-002 77145-004 77145-006 77145-008
(R,R)-(-)-2,5-Hexanediol 98%
(R,R)-(-)-2,5-Hexanediol
Precaución: for Research Use Only


Formula: C₆H₁₄O₂
MW: 118.18
MDL Number: MFCD00142339
CAS Number: 17299-07-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty