Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-tert-Butyl-β-alanine 97%

Supplier: AMBEED, INC
Warning

A624624-100MG A624624-250MG
77145-074EA 39.15 USD InStock
77145-074 77145-076
(R)-3-tert-Butyl-β-alanine 97%
(R)-3-tert-Butyl-β-alanine
Precaución: for Research Use Only


Formula: C₇H₁₅NO₂
MW: 145.2 g/mol
MDL Number: MFCD07784062
CAS Number: 367278-49-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty