Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,1'S,3R,3'R,4S,4'S,6R,6'R)-6,6'-((1S,1'S)-((2,5-Diphenylpyrimidine-4,6-diyl)bis(oxy))bis((6-methoxyquinolin-4-yl)methylene))bis(3-ethylquinuclidine) 97%

Supplier: AMBEED, INC
Warning

A626169-1G A626169-250MG A626169-25G A626169-5G
77145-106EA 84.46 USD InStock
77145-106 77145-108 77145-110 77145-112
(1S,1'S,3R,3'R,4S,4'S,6R,6'R)-6,6'-((1S,1'S)-((2,5-Diphenylpyrimidine-4,6-diyl)bis(oxy))bis((6-methoxyquinolin-4-yl)methylene))bis(3-ethylquinuclidine) 97%
(1S,1'S,3R,3'R,4S,4'S,6R,6'R)-6,6'-((1S,1'S)-((2,5-Diphenylpyrimidine-4,6-diyl)bis(oxy))bis((6-methoxyquinolin-4-yl)methylene))bis(3-ethylquinuclidine)
Precaución: for Research Use Only


Formula: C₅₆H₆₀N₆O₄
MDL Number: MFCD00198108
CAS Number: 149725-81-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty