Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-((tert-Butoxycarbonyl)amino)-2-chlorophenyl)boronic acid 96%

Supplier: AMBEED, INC
Warning

A637569-100MG A637569-1G A637569-250MG
77145-230EA 15.45 USD InStock
77145-230 77145-232 77145-234
(5-((tert-Butoxycarbonyl)amino)-2-chlorophenyl)boronic acid 96%
(5-((tert-Butoxycarbonyl)amino)-2-chlorophenyl)boronic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₅BClNO₄
MDL Number: MFCD22421494
CAS Number: 2246849-87-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty