Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloro-3-(pyridin-4-yl)-2,6-naphthyridine 97%

Supplier: AMBEED, INC
Warning

A646962-100MG A646962-250MG
77145-302EA 372.85 USD InStock
77145-302 77145-304
1-Chloro-3-(pyridin-4-yl)-2,6-naphthyridine 97%
1-Chloro-3-(pyridin-4-yl)-2,6-naphthyridine
Precaución: for Research Use Only


Formula: C₁₃H₈ClN₃
MW: 241.67 g/mol
MDL Number: MFCD18260360
CAS Number: 1071017-49-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty