Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1'-(tert-Butoxycarbonyl)-4-oxospiro[chroman-2,4'-piperidine]-7-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A647132-100MG A647132-50MG
77145-306EA 66.95 USD InStock
77145-306 77145-308
1'-(tert-Butoxycarbonyl)-4-oxospiro[chroman-2,4'-piperidine]-7-carboxylic acid 97%
1'-(tert-Butoxycarbonyl)-4-oxospiro[chroman-2,4'-piperidine]-7-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₉H₂₃NO₆
MDL Number: MFCD11501223
CAS Number: 1086398-20-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty