Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-Pulegone 92%

Supplier: AMBEED, INC
Warning

A656020-25G A656020-5G A656020-500G
77145-344EA 80.34 USD InStock
77145-344 77686-622 77686-621
(R)-(+)-Pulegone 92%
(R)-(+)-Pulegone
Precaución: for Research Use Only


Formula: C₁₀H₁₆O
MDL Number: MFCD00063000
CAS Number: 89-82-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty