Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diallyl maleate 97% stabilized

Supplier: AMBEED, INC
Danger

Stabilization: Stabilised with hydroquinone (HQ)

A665098-100G A665098-25G
77145-426EA 19.57 USD InStock
77145-426 77145-428
Diallyl maleate 97% stabilized
Diallyl maleate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₄
MDL Number: MFCD00010763
CAS Number: 999-21-3
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty