Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Bromo-2-formylphenoxy)acetic acid 97%

Supplier: AMBEED, INC
Warning

A672902-250MG A672902-1G A672902-100MG
77145-496EA 62.83 USD InStock
77145-496 77747-364 77747-363
2-(4-Bromo-2-formylphenoxy)acetic acid 97%
2-(4-Bromo-2-formylphenoxy)acetic acid
Precaución: for Research Use Only


Formula: C₉H₇BrO₄
MDL Number: MFCD02257408
CAS Number: 24589-89-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty