Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dimethylphenoxyacetic acid 98%

Supplier: AMBEED, INC
Danger

A681900-25G A681900-5G
77145-550EA 47.38 USD InStock
77145-550 77145-552
2,6-Dimethylphenoxyacetic acid 98%
2,6-Dimethylphenoxyacetic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
MDL Number: MFCD00156912
CAS Number: 13335-71-2
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty