Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers) 95%

Supplier: AMBEED, INC
Danger

A682349-100G A682349-25G A682349-500G
77145-554EA 8.24 USD InStock
77145-554 77145-556 77145-558
1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers) 95%
1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers)
Precaución: for Research Use Only


Formula: C₈H₁₀O₃
MW: 154.17 g/mol
MDL Number: MFCD00064863
CAS Number: 85-42-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty