Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,2S)-N1,N1,N2,N2-Tetramethyl-1,2-diphenylethane-1,2-diamine 97%

Supplier: AMBEED, INC
Warning

A682369-100MG A682369-1G A682369-250MG
77145-560EA 18.55 USD InStock
77145-560 77145-562 77145-564
(1S,2S)-N1,N1,N2,N2-Tetramethyl-1,2-diphenylethane-1,2-diamine 97%
(1S,2S)-N1,N1,N2,N2-Tetramethyl-1,2-diphenylethane-1,2-diamine
Precaución: for Research Use Only


Formula: C₁₈H₂₄N₂
MDL Number: MFCD06797770
CAS Number: 91361-07-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty