Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,3S)-2-Amino-3-methoxybutanoic acid 95%

Supplier: AMBEED, INC
Warning

A685848-100MG A685848-1G A685848-250MG A685848-5G
77145-610EA 101.96 USD InStock
77145-610 77145-612 77145-614 77145-616
(2S,3S)-2-Amino-3-methoxybutanoic acid 95%
(2S,3S)-2-Amino-3-methoxybutanoic acid
Precaución: for Research Use Only


Formula: C₅H₁₁NO₃
MDL Number: MFCD00142985
CAS Number: 104195-80-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty