Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-Bromoethyl)benzene 97%

Supplier: AMBEED, INC
Warning

A687693-25G A687693-500G A687693-5G
77145-624EA 39.14 USD InStock
77145-624 77145-626 77145-628
(1-Bromoethyl)benzene 97%
(1-Bromoethyl)benzene
Precaución: for Research Use Only


Formula: C₆H₅CHBrCH₃
MW: 185.06 g/mol
Boiling Pt: 203
Melting Pt: 50
MDL Number: MFCD00000139
CAS Number: 585-71-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty