Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluorobenzo[d]oxazole-2(3H)-thione 97%

Supplier: AMBEED, INC
Danger

A697170-100MG A697170-1G A697170-250MG
77145-748EA 131.84 USD InStock
77145-748 77145-750 77145-752
4-Fluorobenzo[d]oxazole-2(3H)-thione 97%
4-Fluorobenzo[d]oxazole-2(3H)-thione
Precaución: for Research Use Only


Formula: C₇H₄FNOS
MDL Number: MFCD28665687
CAS Number: 1163123-40-7
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty