Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Famotidine 97%

Supplier: AMBEED, INC
Warning

A701177-100G A701177-1G A701177-25G A701177-100MG A701177-50MG A701177-5G
77145-780EA 329.6 USD InStock
77145-780 77145-782 77145-784 77686-686 77686-687 77686-688
Famotidine 97%
Famotidine
Precaución: for Research Use Only


Formula: C₈H₁₅N₇O₂S₃
MDL Number: MFCD00079297
CAS Number: 76824-35-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty