Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-methoxyisonicotinic acid 95%

Supplier: AMBEED, INC
Warning

A702743-10G A702743-1G A702743-250MG A702743-5G
77145-794EA 128.75 USD InStock
77145-794 77145-796 77145-798 77145-800
2-Chloro-6-methoxyisonicotinic acid 95%
2-Chloro-6-methoxyisonicotinic acid
Precaución: for Research Use Only


Formula: C₇H₆ClNO₃
MW: 187.58
MDL Number: MFCD00173928
CAS Number: 15855-06-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty