Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,4S)-4-Hydroxycyclopent-2-en-1-yl acetate 95%

Supplier: AMBEED, INC
Warning

A707073-1G A707073-250MG A707073-100MG
77145-854EA 183.34 USD InStock
77145-854 77145-856 77686-693
(1R,4S)-4-Hydroxycyclopent-2-en-1-yl acetate 95%
(1R,4S)-4-Hydroxycyclopent-2-en-1-yl acetate
Precaución: for Research Use Only


Formula: C₇H₁₀O₃
MDL Number: MFCD00210003
CAS Number: 60410-16-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty