Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-fluoroanisole 98%

Supplier: AMBEED, INC
Warning

A712311-100G A712311-1G A712311-250MG A712311-25G A712311-5G
77145-892EA 363.59 USD InStock
77145-892 77145-894 77145-896 77145-898 77145-900
3-Chloro-5-fluoroanisole 98%
3-Chloro-5-fluoroanisole
Precaución: for Research Use Only


Formula: C₇H₆ClFO
MDL Number: MFCD00070770
CAS Number: 202925-08-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty