Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl pyrrolo[2,1-f][1,2,4]triazine-7-carboxylate 97%

Supplier: AMBEED, INC
Warning

A716772-100MG A716772-250MG
77145-936EA 109.18 USD InStock
77145-936 77145-938
Ethyl pyrrolo[2,1-f][1,2,4]triazine-7-carboxylate 97%
Ethyl pyrrolo[2,1-f][1,2,4]triazine-7-carboxylate
Precaución: for Research Use Only


Formula: C₉H₉N₃O₂
MDL Number: MFCD21607356
CAS Number: 1356016-44-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty